Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tributyltin chloride 95% tech.

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tri-n-butyltin chlorideChlorotributylstannaneTBTC

AC13935-1000 AC13935-5000
TS13935-1000EA 60 USD InStock
TS13935-1000 TS13935-5000
Tributyltin chloride 95% tech.
Tributyltin chloride


Formula: C₁₂H₂₇ClSn
MW: 325.51 g/mol
Boiling Pt: 171…173 °C (25 mmHg)
Melting Pt: –9 °C
Density: 1.2 g/cm³
Flash Pt: >112 °C
MDL Number: MFCD00000521
CAS Number: 1461-22-9
EINECS: 215-958-7
UN: 2788
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4940 (20°C, 589 nm)
Titration Argentometric ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty