Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl formate, extra dry 98+%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Formic acid ethyl ester

AC42924-1000
TS42924-1000EA 49.77 USD InStock
TS42924-1000
Ethyl formate, extra dry 98+%, AcroSeal™
Ethyl formate


Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 54 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.917 g/cm³
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003294
CAS Number: 109-94-4
UN: 1190
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.0 %
UV 1 cm cell vs HPLC-grade water
UV at 240 nm A: ≤0.301
UV at 255 nm A: ≤0.097
UV at 280 nm A: ≤0.015
Residue after evaporation ≤0.0003 %
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0002 meq/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty