Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrimethoxysilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethoxy(methyl)silane

AC25054-0500 AC25054-2500 AC25054-0050 AC25054-0010
TS25054-2500EA 36 USD InStock
TS25054-2500 TS25054-0500 TS25054-0050 TS25054-0010
Methyltrimethoxysilane 97%
Methyltrimethoxysilane


Formula: C₄H₁₂O₃Si
Boiling Pt: 102 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.95 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008342
CAS Number: 1185-55-3
EINECS: 214-685-0
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3695 to 1.3715 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty