Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Neodymium trichloride, anhydrous 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Neodymium (III) chloride

AC31507-0050
TS31507-0050EA 95 USD InStock
TS31507-0050
Neodymium trichloride, anhydrous 99.9% (trace metals basis)
Neodymium trichloride


Formula: Cl₃Nd
MW: 250.6 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 784 °C
MDL Number: MFCD00011132
CAS Number: 10024-93-8
EINECS: 233-031-5
Order Now
Specification Test Results
Appearance White to light green Powder
Total conc. of imp.as REO ≤0.1 %
Neodymium (Nd) 55.8 to 59.3 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty