Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) ethanolate (33 - 35% TiO₂)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Titanium (IV) ethoxideTitanium tetraethanolateTetraethyl orthotitanate

AC22376-2500 AC22376-0500 AC22376-0010
TS22376-0500EA 66 USD InStock
TS22376-0500 TS22376-0010 TS22376-2500
Titanium(IV) ethanolate (33 - 35% TiO₂)
Titanium(IV) ethanolate


Formula: C₈H₂₀O₄Ti
MW: 228.12 g/mol
Boiling Pt: 150…152 °C (10 mmHg)
Melting Pt: –40 °C
Density: 1.09 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009071
CAS Number: 3087-36-3
EINECS: 221-410-8
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5070 (20°C, 589 nm)
Titration Complexometric 33 to 35 % (TiO2)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty