Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Tetrylammonium bromide

AC16107-0050 AC16107-2500 AC16107-0010
TS16107-2500EA 64.34 USD InStock
TS16107-2500 TS16107-0010 TS16107-0050
Tetraethylammonium bromide 98%
Tetraethylammonium bromide


Formula: C₈H₂₀BrN
MW: 210.16 g/mol
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011825
CAS Number: 71-91-0
EINECS: 200-769-4
Merck Index: 12,09340
Order Now
Specification Test Results
Appearance White to cream Crystalline powder or crystals or solid
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Loss on drying ≤2 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty