Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethyltin 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tin tetramethylTetramethylstannane

AC16398-0500 AC16398-0100
TS16398-0500EA 352 USD InStock
TS16398-0500 TS16398-0100
Tetramethyltin 98%
Tetramethyltin


Formula: C₄H₁₂Sn
MW: 178.85 g/mol
Boiling Pt: 75 °C (1013 hPa)
Melting Pt: –54 °C
Density: 1.29 g/cm³
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008278
CAS Number: 594-27-4
EINECS: 209-833-6
UN: 3384
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty