Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ytterbium(III) oxide 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals

AC26430-0100
TS26430-0100EA 131 USD InStock
TS26430-0100
Ytterbium(III) oxide 99.99% (trace metals basis)
Ytterbium (III) oxide


Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2346 °C
Storage Temperature: Ambient
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤100 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty