Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ruthenium(IV) oxide hydrate (> 54% Ru)

Supplier: Thermo Scientific Chemicals
Warning

AC19743-0250 AC19743-0010 AC19743-0050
TS19743-0050EA 310 USD InStock
TS19743-0050 TS19743-0250 TS19743-0010
Ruthenium(IV) oxide hydrate (> 54% Ru)
Ruthenium (IV) oxide hydrate


Formula: O₂Ru·H₂O
MW: 151.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149846
CAS Number: 32740-79-7
EINECS: 234-840-6
Order Now
Specification Test Results
Appearance Black Powder
Assay ≥54 % (Ru)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty