Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylaluminium 0.6 M in heptane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triethylaluminum

AC38117-1000 AC38117-8000
TS38117-8000EA 436 USD InStock
TS38117-8000 TS38117-1000
Triethylaluminium 0.6 M in heptane, AcroSeal™
Triethylaluminium


Formula: C₆H₁₅Al
MW: 114.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3399
ADR: 4.3,I
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 9.5 to 10.5 wt % (on Al basis)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty