Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyltin dichloride 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dichlorodimethylstannane

AC34782-0250 AC34782-0050 AC34782-1000
TS34782-0250EA 156 USD InStock
TS34782-0250 TS34782-1000 TS34782-0050
Dimethyltin dichloride 95%
Dimethyltin dichloride


Formula: C₂H₆Cl₂Sn
MW: 219.69 g/mol
Boiling Pt: 188…190 °C (760 mmHg)
Melting Pt: 103…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 103 to 110 °C
Titration Argentometric ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty