Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lutetium(III) oxide 99.99% (trace metals basis) -325 mesh

Supplier: Thermo Scientific Chemicals
Synonyms: Lutetium oxideDilutetium trioxide

AC19340-0010
TS19340-0010EA 108 USD InStock
TS19340-0010
Lutetium(III) oxide 99.99% (trace metals basis) -325 mesh
Lutetium (III) oxide


Formula: Lu₂O₃
MW: 397.93 g/mol
Melting Pt: 2510 °C
Density: 9.42 g/cm³
MDL Number: MFCD00011100
CAS Number: 12032-20-1
EINECS: 234-764-3
Order Now
Specification Test Results
Appearance White Powder
Assay ≥99.94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty