Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver cyanide 99% for analysis

Supplier: Thermo Scientific Chemicals
Danger

AC18023-0100 AC18023-0500
TS18023-0500EA 223 USD InStock
TS18023-0500 TS18023-0100
Silver cyanide 99% for analysis
Silver cyanide


Formula: AgCN
MW: 133.89 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003409
CAS Number: 506-64-9
EINECS: 208-048-6
UN: 1684
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White Powder
Assay ≥98.5 %
Aluminium (Al) ≤10 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤30 ppm
Lead (Pb) ≤10 ppm
Sodium (Na) ≤500 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty