Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium acetate xhydrate 99% for analysis

Supplier: Thermo Scientific Chemicals
Synonyms: Calcium acetate hydrateAcetic acid calcium salt hydrateCalcium di(acetate) hydrate

AC44786-0010
TS44786-0010EA 110.61 USD InStock
TS44786-0010
Calcium acetate xhydrate 99% for analysis
Calcium acetate xhydrate


Formula: (H₃CCOO)₂Ca·nH₂O
MW: 176.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150019
CAS Number: 114460-21-8
Order Now
Specification Test Results
Appearance White Powder or granules
Infrared spectrum Conforms
Titration Complexometric ≥98.5 % (on dried substance)
Water ≤10 %
pH 6.0 to 9.0 (10 % in water)
Chloride (Cl) ≤0.05 %
Sulfate (SO4) ≤0.1 %
Arsenic (As) ≤3 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤2 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.1 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty