Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexachloroplatinic(IV) acid hydrate 99.995% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hydrogen hexachloroplatinate (IV) hydrateDihydrogen hexachloroplatinate (IV) hydrateHydrogen hexachloroplatinate(IV) hydrateChloroplatinic acid hydrateHydrogen hexachloroplatinate hydrate

AC43720-0050 AC43720-0010
TS43720-0010EA 159.95 USD InStock
TS43720-0010 TS43720-0050
Hexachloroplatinic(IV) acid hydrate 99.995% (trace metals basis)
Hexachloroplatinic (IV) acid hydrate


Formula: H₂Cl₆Pt·H₂O
MW: 427.83 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149909
CAS Number: 26023-84-7
UN: 2507
ADR: 8,III
Order Now
Specification Test Results
Appearance Orange to orange-brown Powder or crystals and/or chunks
Assay ≥37.0 % (Pt)
Total trace metal impurities ≤50 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty