Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium(III) sulfate octahydrate 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dicerium trisulphate octahydrateDicerium trisulfate octahydrate

AC19279-0500 AC19279-2500
TS19279-2500EA 739 USD InStock
TS19279-2500 TS19279-0500
Cerium(III) sulfate octahydrate 99.99% (trace metals basis)
Cerium (III) sulphate octahydrate


Formula: Ce₂O₁₂S₃·8H₂O
MW: 712.54 g/mol
MDL Number: MFCD00149641
CAS Number: 10450-59-6
EINECS: 236-644-6
Order Now
Specification Test Results
Appearance White to off-white Crystals and/or chunks
Assay 38.0 to 40.5 % (Ce)
Total trace metal impurities ≤0.01 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty