Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(II) perchlorate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

AC20136-5000 AC20136-1000 AC20136-0050
TS20136-0050EA 38.9 USD InStock
TS20136-0050 TS20136-5000 TS20136-1000
Copper(II) perchlorate hexahydrate 98%
Copper (II) perchlorate hexahydrate


Formula: Cl₂CuO₈·6H₂O
MW: 370.54 g/mol
Melting Pt: 82 °C
Density: 2.22 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149666
CAS Number: 10294-46-9
EINECS: 237-391-4
UN: 1481
ADR: 5.1,II
Order Now
Specification Test Results
Appearance Blue Adhering crystals
Titration Iodometric ≥97.5 %
pH 4 to 7 (5 % solution)
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤50 ppm
Insoluble matter ≤0.01 % (in water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty