Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate sesquihydrate 98.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

AC34720-2500 AC34720-0010 AC34720-0025
TS34720-2500EA 57 USD InStock
TS34720-2500 TS34720-0025 TS34720-0010
Potassium carbonate sesquihydrate 98.5% ACS
Potassium carbonate sesquihydrate


Formula: CK₂O₃·1.5H₂O
MW: 165.23 g/mol
Melting Pt: 891 °C
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3
Order Now
Specification Test Results
Appearance White Crystalline powder
Titration with HCl 98.5 to 101.0 %
Chloride (Cl) ≤0.003 %
Phosphate (PO4) ≤0.001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Silicon dioxide (SiO2) ≤0.005 %
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %
Loss on heating 14.0 % to 16.5 % (285°C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty