Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium metavanadate ACS

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ammonium vanadium oxideAmmonium monovanadateAmmonium vanadium trioxide

AC44726-1000 AC44726-5000 AC44726-0250
TS44726-1000EA 64.95 USD InStock
TS44726-1000 TS44726-5000 TS44726-0250
Ammonium metavanadate ACS
Ammonium metavanadate


Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
UN: 2859
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to yellow to tan Powder
Solubility (in Ammonium hydroxide) Passes test
Titration with KMnO4 ≥99.0 %
Chloride (Cl) ≤0.2 %
Carbonate (CO3) (Limit about 0.3 %) Passes test
Sulfate (SO4) ≤0.05 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty