Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) acetylacetonate 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel acetylacetonateBis(2,4-pentanedionato-O,O')nickel (II)Nickel (II) 2,4-pentanedionateNickel (II) bis(2,4-pentanedionato-O,O')

AC12826-1000 AC12826-0250 AC12826-0050
TS12826-0050EA 43.78 USD InStock
TS12826-0050 TS12826-0250 TS12826-1000
Nickel(II) acetylacetonate 96%
Nickel (II) acetylacetonate


Formula: C₁₀H₁₄NiO₄
MW: 256.91 g/mol
Boiling Pt: 220 °C (11 mmHg)
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000024
CAS Number: 3264-82-2
EINECS: 221-875-7
Order Now
Specification Test Results
Appearance Green Powder
Infrared spectrum Conforms
Titration Complexometric ≥95 %
Chloride (Cl) ≤5 ppm
Phosphate (PO4) ≤5 ppm
Calcium (Ca) ≤10 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Potassium (K) ≤10 ppm
Sodium (Na) ≤20 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty