Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) acetylacetonate (35% Pd)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Palladium (II) bis(2,4-pentanedionato-O,O')Palladium (II) 2,4-pentanedionateBis(2,4-pentanedionato-O,O')palladium (II)Palladium(II) 2,4-pentanedionate

AC19758-0050 AC19758-0010
TS19758-0050EA 541 USD InStock
TS19758-0050 TS19758-0010
Palladium(II) acetylacetonate (35% Pd)
Palladium (II) acetylacetonate


Formula: C₁₀H₁₄O₄Pd
MW: 304.64 g/mol
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8
Order Now
Specification Test Results
Appearance Yellow to green Powder
Infrared spectrum Conforms
Assay 33.9 to 35.9 % (Pd)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty