Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dichloroacetate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 2,2-dichloroacetate

AC33828-0500 AC33828-0100
TS33828-0100EA 67 USD InStock
TS33828-0100 TS33828-0500
Sodium dichloroacetate 98%
Sodium dichloroacetate


Formula: C₂HCl₂NaO₂
MW: 150.92 g/mol
Melting Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00070489
CAS Number: 2156-56-1
EINECS: 218-461-3
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty