Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hexachloroplatinate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hexachloroplatinate(IV) hexahydrateSodium hexachloroplatinate (IV) hexahydrate

AC31675-0010
TS31675-0010EA 140 USD InStock
TS31675-0010
Disodium hexachloroplatinate hexahydrate 98%
Disodium hexachloroplatinate hexahydrate


Formula: Cl₆Na₂Pt·6H₂O
MW: 561.87 g/mol
MDL Number: MFCD00149172
CAS Number: 19583-77-8
EINECS: 240-983-5
Order Now
Specification Test Results
Appearance Orange Crystalline solid
Assay ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty