Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium perborate tetrahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium peroxometaborate tetrahydrate

AC22320-0250 AC22320-2500 AC22320-0010 AC22320-0050
TS22320-0050EA 361 USD InStock
TS22320-0050 TS22320-0010 TS22320-0250 TS22320-2500
Sodium perborate tetrahydrate 97%
Sodium perborate tetrahydrate


Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
Order Now
Specification Test Results
Appearance White Crystalline powder
pH 10.1 to 10.6 (1 % solution)
Titration with KMnO4 ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty