Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodotrimethylsilane 95-97% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethylsilyl iodideTrimethyliodosilane

AC18749-0050 AC18749-0250 AC18749-2500 AC18749-1000
TS18749-0050EA 63 USD InStock
TS18749-0050 TS18749-0250 TS18749-1000 TS18749-2500
Iodotrimethylsilane 95-97% stabilized
Iodotrimethylsilane


Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 106 °C (760 mmHg)
Density: 1.4 g/cm³
Flash Pt: –31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to red Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4800 (20°C, 589 nm)
Titration Argentometric ≥95 % (Ex I)
Stabilizer copper

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty