Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylsulfonium iodide 98%

Supplier: Thermo Scientific Chemicals

AC14025-0250 AC14025-1000 AC14025-5000
TS14025-1000EA 115 USD InStock
TS14025-1000 TS14025-5000 TS14025-0250
Trimethylsulfonium iodide 98%
Trimethylsulphonium iodide


Formula: C₃H₉IS
MW: 204.07 g/mol
Melting Pt: 215…220 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4
Order Now
Specification Test Results
Appearance White to yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty