Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum(III) chloride hydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

AC41301-0500
TS41301-0500EA 59 USD InStock
TS41301-0500
Lanthanum(III) chloride hydrate ACS
Lanthanum (III) chloride hydrate


Formula: Cl₃La·H₂O
MW: 263.28 g/mol
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149753
CAS Number: 20211-76-1
EINECS: 233-237-5
UN: 1759
Order Now
Specification Test Results
Appearance White Adhering crystalline powder or crystals
Assay 64.5 to 70.0 % (LaCl3)
Calcium (Ca) ≤10 ppm
Magnesium (Mg) ≤10 ppm
Insoluble matter ≤0.01 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty