Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Chloromethyl)dimethylchlorosilane 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: CMDMCSChloro(chloromethyl)dimethylsilane

AC43402-1000
TS43402-1000EA 126 USD InStock
TS43402-1000
(Chloromethyl)dimethylchlorosilane 98%, AcroSeal™
(Chloromethyl)dimethylchlorosilane


Formula: C₃H₈Cl₂Si
Boiling Pt: 114 °C (1013hPa)
Density: 1.08 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000875
CAS Number: 1719-57-9
UN: 2985
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4360 to 1.4380 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty