Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorodimethylvinylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DimethylvinylchlorosilaneVinyldimethylchlorosilane

AC33673-0250 AC33673-0050
TS33673-0050EA 40 USD InStock
TS33673-0050 TS33673-0250
Chlorodimethylvinylsilane 97%
Chlorodimethylvinylsilane


Formula: C₄H₉ClSi
Flash Pt: –4 °C
MDL Number: MFCD00018090
CAS Number: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty