Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(diphenyl)silane 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: DichlorodiphenylsilaneDiphenyldichlorosilane

AC11334-5000 AC11334-1000
TS11334-5000EA 225 USD InStock
TS11334-5000 TS11334-1000
Dichloro(diphenyl)silane 97%
Dichloro(diphenyl)silane


Formula: C₁₂H₁₀Cl₂Si
MW: 253.2 g/mol
Boiling Pt: 305 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.204 g/cm³
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00000489
CAS Number: 80-10-4
EINECS: 201-251-0
UN: 1769
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5765 to 1.5815 (20°C, 589 nm)
GC ≥96.0 %
Absorption Ratio P-15/P+15 1.1 to 1.32
BSC Certification Passes

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty