Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butylchlorodiphenylsilane 98%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyldiphenylchlorosilanetert-Butyl-diphenylchlorosilane

AC43092-1000
TS43092-1000EA 240 USD InStock
TS43092-1000
tert-Butylchlorodiphenylsilane 98%, AcroSeal™
tert-Butylchlorodiphenylsilane


Formula: C₁₆H₁₉ClSi
MW: 274.87 g/mol
Boiling Pt: 135 °C (.7 mmHg)
Density: 1.06 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00000497
CAS Number: 58479-61-1
EINECS: 261-282-0
UN: 2987
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5665 to 1.5685 (20°C, 589 nm)
GC ≥97.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty