Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Picolyl chloride hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(Chloromethyl)pyridine hydrochloride

AC17212-0250 AC17212-1000 AC17212-0050
TS17212-0050EA 38 USD InStock
TS17212-0050 TS17212-1000 TS17212-0250
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride


Formula: C₆H₇Cl₂N
MW: 164.03 g/mol
Melting Pt: 119…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00012811
CAS Number: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Appearance Yellow-green to light brown Crystalline mass
Infrared spectrum Conforms
Melting point 119 to 125 °C
Titration with HClO4 ≥97.5 %
Specific optical rotation -115° to -130° (20 °C, 589 nm) (c=0.8 in EtOH)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty