Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(methyl)(phenyl)silane 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methylphenyldichlorosilane

AC14738-1000 AC14738-5000 AC14738-0100
TS14738-1000EA 47 USD InStock
TS14738-1000 TS14738-0100 TS14738-5000
Dichloro(methyl)(phenyl)silane 98%
Dichloro(methyl)(phenyl)silane


Formula: C₇H₈Cl₂Si
MW: 191.13 g/mol
Density: 1.176 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000490
CAS Number: 149-74-6
EINECS: 205-746-2
UN: 2437
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5180 to 1.5200 (20°C, 589 nm)
GC ≥97.5 %
Titration excess HCl 40.0 to 43.5 % (MgO)
Arsenic (As) ≤4 ppm
Calcium (Ca) ≤0.45 %
Iron (Fe) ≤0.02 %
Acid insoluble substances ≤0.05 %
Soluble salts ≤1.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty