Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibutyltin dilaurate 94%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Di-n-butyltin dilaurate

AC38269-0050
TS38269-0050EA 45 USD InStock
TS38269-0050
Dibutyltin dilaurate 94%
Dibutyltin dilaurate


Formula: C₃₂H₆₄O₄Sn
MW: 631.57 g/mol
Boiling Pt: 205 °C (12 mmHg)
Melting Pt: 16…18 °C
Density: 1.05 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00008963
CAS Number: 77-58-7
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4690 to 1.4730 (20°C, 589 nm)
Tin (Sn) 17.5 to 19.9 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty