Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloro(dimethyl)(phenyl)silane 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorodimethylphenylsilane

AC15107-1000 AC15107-0050 AC15107-0250
TS15107-0050EA 40 USD InStock
TS15107-0050 TS15107-0250 TS15107-1000
Chloro(dimethyl)(phenyl)silane 95%
Chloro(dimethyl)(phenyl)silane


Formula: C₈H₁₁ClSi
MW: 170.71 g/mol
Boiling Pt: 80…84 °C (16 mmHg)
Density: 1.032 g/cm³
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00000499
CAS Number: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5060 to 1.5100 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty