Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylarsine oxide 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: OxophenylarsineArzenePAO

AC18035-0010 AC18035-0250 AC18035-0050
TS18035-0250EA 441 USD InStock
TS18035-0250 TS18035-0050 TS18035-0010
Phenylarsine oxide 97%
Phenylarsine oxide


Formula: C₆H₅AsO
MW: 168.02 g/mol
Melting Pt: 145…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00001990
CAS Number: 637-03-6
EINECS: 211-275-3
UN: 3465
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Assay ≥96.0 %
HPLC ≥98.5 %
Optical rotation +24° to +28° (1 % in DMF/methanol)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty