Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethynyltributyltin 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethynyltri-n-butylstannaneTributylethynylstannane

AC38918-0025
TS38918-0025EA 144 USD InStock
TS38918-0025
Ethynyltributyltin 95%
Ethynyltributyltin


Formula: C₁₄H₂₈Sn
MW: 315.09
Boiling Pt: 70 °C (.2 mmHg)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00009420
CAS Number: 994-89-8
UN: 2788
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4770 (20°C, 589 nm)
GC ≥94.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty