Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(Pentafluoroethyl)trimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

AC45930-0050 AC45930-0010 AC45930-0250
TS45930-0050EA 413 USD InStock
TS45930-0050 TS45930-0250 TS45930-0010
(Pentafluoroethyl)trimethylsilane 97%
(Pentafluoroethyl)trimethylsilane


Formula: C₅H₉F₅Si
Boiling Pt: 70 °C
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD04116436
CAS Number: 124898-13-1
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to almost colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3240 to 1.3260 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty