Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluenesulfinic Acid Sodium Salt, anhydrous 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 4-methylbenzenesulphinateSodium-p-toluenesulphinateSodiumtoluene-4-sulphinate4-Methylbenzenesulphinic acid sodium saltSodium 4-methylbenzenesulfinate4-Methylbenzenesulfinic acid sodium saltSodiumtoluene-4-sulfinateSodium-p-toluenesulfinate

AC43453-0250 AC43453-1000
TS43453-1000EA 109 USD InStock
TS43453-1000 TS43453-0250
p-Toluenesulfinic Acid Sodium Salt, anhydrous 97%
p-Toluenesulfinic Acid Sodium Salt


Formula: C₇H₇NaO₂S
MW: 178.19 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013136
CAS Number: 824-79-3
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤3.00 %
HPLC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty