Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium tert-butoxide 93+%

Supplier: Thermo Scientific Chemicals
Warning

AC39047-0100 AC39047-2500 AC39047-0500
TS39047-0500EA 113 USD InStock
TS39047-0500 TS39047-2500 TS39047-0100
Magnesium tert-butoxide 93+%
Magnesium tert-butoxide


Formula: C₈H₁₈MgO₂
MW: 170.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788761
CAS Number: 32149-57-8
UN: 3205
ADR: 4.2,II
Order Now
Specification Test Results
Appearance White to light gray Powder
Titration with HCl ≥93.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty