Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tebbe Reagent 0.5 M in toluene

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bis(cyclopentadienyl-mu-chloro(dimethylaluminium-mu-methylenetitaniumµ-Chloro-µ-methylenebis(cyclopentadienyl)titaniumdimethylaluminum

AC36897-0250 AC36897-1000
TS36897-0250EA 244 USD InStock
TS36897-0250 TS36897-1000
Tebbe Reagent 0.5 M in toluene
Tebbe Reagent


Formula: C₁₃H₁₈AlClTi
MW: 284.62 g/mol
Density: 0.927 g/cm³
Flash Pt: 4 °C
Storage Temperature: Freezer
MDL Number: MFCD00151575
CAS Number: 67719-69-1
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Dark red to purple Solution
Strength 0.40 to 0.60 M

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty