Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Bis(t-butylcyclopentadienyl)titanium dichloride

AC34786-0010
TS34786-0010EA 93 USD InStock
TS34786-0010
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride


Formula: C₁₈H₂₆Cl₂Ti
MW: 361.172 g/mol
Melting Pt: 233 °C
Storage Temperature: Ambient
MDL Number: MFCD01073774
CAS Number: 79269-71-9
Order Now
Specification Test Results
Appearance Bordeaux Crystals
Infrared spectrum Conforms

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty