Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Palladium 5% on barium sulfate hydrogenation catalyst (oxidised form), unreduced

Supplier: Thermo Scientific Chemicals

AC19506-0100 AC19506-0500
TS19506-0100EA 159 USD InStock
TS19506-0100 TS19506-0500
Palladium 5% on barium sulfate hydrogenation catalyst (oxidised form), unreduced
Palladium


Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Order Now
Specification Test Results
Appearance Brown Powder
Assay 4.50 to 5.50 % (Pd)
Water ≤5.0 % (K.F.)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty