Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium(II) nitrate hydrate

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Palladium dinitrate hydratePalladous nitrate hydrate

AC38991-0250 AC38991-0010 AC38991-0050
TS38991-0010EA 162.85 USD InStock
TS38991-0010 TS38991-0250 TS38991-0050
Palladium(II) nitrate hydrate
Palladium(II) nitrate hydrate


Formula: N₂O₆Pd·xH₂O
MW: 230.43 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149818
CAS Number: 207596-32-5
UN: 3085
ADR: 5.1,II
Order Now
Specification Test Results
Appearance Brown to brown-black Crystals
Palladium (Pd) 39 to 46 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty