Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide basic Brockmann Grade I 50-200 µm, 90A for chromatography

Supplier: Thermo Scientific Chemicals

AC46335-0010
TS46335-0010EA 109.35 USD InStock
TS46335-0010
Aluminium oxide basic Brockmann Grade I 50-200 µm, 90A for chromatography
Aluminium oxide basic


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2977 °C (760 mmHg)
Melting Pt: 2030 °C
Density: Mar-97
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now
Specification Test Results
Appearance White to almost white Crystalline powder
Particle size ≤10 % (< 50 um)
Particle size ≤5 % (> 200 um)
Specific surface area 90 to 170 m2/g,N2 adsorp
pH 8.5 to 10.5 (10 % suspension in water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty