Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid 99.5%, pure

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

12404-0010 12404-0025
TS12404-0010EA 68 USD InStock
TS12404-0010 TS12404-0025
Acetic acid 99.5%, pure
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 16…16.5 °C
Density: 1.048 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3700 to 1.3730 (20 °C, 589 nm)
Color scale ≤10 APHA
Titration with NaOH ≥99.4 %
Appearance of solution (EP) Passes test
Identification Passes test (EP, USP)
Related substances ≤0.4 % (sum of impurities A and B)
Related substances ≤0.2 % (impurity C)
Related substances ≤0.15 % (impurity D)
Related substances ≤0.10 % (unspecified impurities)
Related substances ≤0.5 % (total impurities)
Specific optical rotation +52.5° to +53.3° (20 °C, 589 nm) (C=10,H2O/dilute NH3) (Anhydrous substance)
Acidity (USP) Passes test
Color of solution (USP) Passes test
Chloride (Cl) ≤0.018 %
Sulfate (SO4) ≤0.025 %
Arsenic (As) ≤1 µg/g
Soluble starch, sulfite ≤15 ppm (EP)
Soluble starch, sulfite (USP) Passes test
Dextrin (EP, USP) Passes test
Conductivity ≤20 µS/cm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty