Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Isopropylamine 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Aminopropane

14892-0100 14892-0250 14892-0010 14892-0025
TS14892-0010EA 50.29 USD InStock
TS14892-0010 TS14892-0100 TS14892-0250 TS14892-0025
Isopropylamine 99%
Isopropylamine


Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 33…34 °C (760 mmHg)
Melting Pt: –101 °C
Density: 0.69 g/cm³
Flash Pt: –37 °C
Storage Temperature: Ambient
MDL Number: MFCD00008082
CAS Number: 75-31-0
EINECS: 200-860-9
UN: 1221
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3725 to 1.3755 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤50 APHA

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty