Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Octane 97%, pure

Supplier: Thermo Scientific Chemicals
Danger

32595-0025 32595-0010
TS32595-0010EA 209.07 USD InStock
TS32595-0010 TS32595-0025
n-Octane 97%, pure
n-Octane


Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 125…127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.708 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,II
Merck Index: 12,06847
https://us-legacy.vwr.com/stibo/web/std.lang.all/79/71/737971.jpg
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3965 to 1.3985 (20 °C, 589 nm)
GC ≥96.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty