Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethylhexanal 96%

Supplier: Thermo Scientific Chemicals
Warning

41009-0010 41009-2500
TS41009-0010EA 201 USD InStock
TS41009-0010 TS41009-2500
2-Ethylhexanal 96%
2-Ethylhexanal


Formula: C₈H₁₆O
MW: 128.21 g/mol
Boiling Pt: 163 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0.8195 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00006987
CAS Number: 123-05-7
EINECS: 204-596-5
UN: 1191
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4140 to 1.4180 (20°C, 589 nm)
GC ≥95 %
Specific gravity (20/20°C) 0.819 to 0.824
Acid value ≤8 mg KOH/g

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty