Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium iodide, anhydrous 99%, pure

Supplier: Thermo Scientific Chemicals
Warning

20359-2500 AC20359-0500
TS20359-0500EA 238.43 USD InStock
TS20359-0500 TS20359-2500
Lithium iodide, anhydrous 99%, pure
Lithium iodide


Formula: ILi
MW: 133.85 g/mol
Boiling Pt: 1177 °C (1013 hPa)
Melting Pt: 446 °C
Storage Temperature: Ambient
MDL Number: MFCD00011092
CAS Number: 10377-51-2
EINECS: 233-822-5
Order Now
Specification Test Results
Appearance White to yellow to beige Powder or crystals or granules
Titration Argentometric ≥98.5 % (on anhydrous substance)
Water ≤1 % (K.F.)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty